4-Chloro-2-iodobenzoic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C2211-D.5 C2211-D.25
XXXXC2211-D.5EA 0 GBP
XXXXC2211-D.5 XXXXC2211-D.25
4-Chloro-2-iodobenzoic acid
4-Chloro-2-iodobenzoic acid
Formula: C₇H₄ClIO₂
MW: 282.46 g/mol
MDL Number: MFCD00060676
CAS Number: 13421-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR