Vitamin A acetate

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: all-trans-Retinyl acetate(2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate

46958.EA 55.9 GBP
46958.
Vitamin A acetate
all-trans-Retinyl acetate
Formula: C₂₂H₃₂O₂
MW: 328.49 g/mol
MDL Number: MFCD00019413
CAS Number: 127-47-9
EINECS: 204-844-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR