Retinol all trans, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: all-trans-Retinol, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol, Vitamin A alcohol

R7632-100MG R7632-1G R7632-250MG R7632-25MG R7632-500MG
SIALR7632-100MGEA 111 GBP
SIALR7632-100MG SIALR7632-1G SIALR7632-250MG SIALR7632-25MG SIALR7632-500MG
Retinol all trans, Sigma-Aldrich®
Retinol all trans
Formula: C₂₀H₃₀O
MW: 286.46 g/mol
MDL Number: MFCD00001552
CAS Number: 68-26-8
EINECS: 200-683-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR