2,6-Dimethylpiperazine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

D179809-1G D179809-25G D179809-5G
SIALD179809-1GEA 50.9 GBP
SIALD179809-1G SIALD179809-25G SIALD179809-5G
2,6-Dimethylpiperazine, Sigma-Aldrich®
2,6-Dimethylpiperazine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD00005956
CAS Number: 108-49-6
EINECS: 203-588-9
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR