TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid

93359-100G 93359-25G
SIAL93359-100GEA 849 GBP
SIAL93359-100G SIAL93359-25G
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Formula: C₆H₁₅NO₆S
MW: 229.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007532
CAS Number: 7365-44-8
EINECS: 230-906-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR