1-(2,4,6-Trimethylbenzyl)piperazine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

651680-1G 651680-5G
SIAL651680-1GEA 66.9 GBP
SIAL651680-1G SIAL651680-5G
1-(2,4,6-Trimethylbenzyl)piperazine, Sigma-Aldrich®
1-(2,4,6-Trimethylbenzyl)piperazine
Formula: C₁₄H₂₂N₂
MW: 218.34 g/mol
MDL Number: MFCD03094587
CAS Number: 41717-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR