Terbium(III) trifluoromethanesulphonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

425729-1G 425729-5G
SIAL425729-1GEA 90 GBP
SIAL425729-1G SIAL425729-5G
Terbium(III) trifluoromethanesulphonate, Sigma-Aldrich®
Terbium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Tb
MW: 606.14 g/mol
MDL Number: MFCD00192461
CAS Number: 148980-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR