4-Nitrophthalic anhydride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Nitroisobenzofuran-1,3-dione

M-Clarity™ quality level = MQ200

238201-25G 238201-5G
SIAL238201-25GEA 249 GBP
SIAL238201-25G SIAL238201-5G
4-Nitrophthalic anhydride, Sigma-Aldrich®
4-Nitrophthalic anhydride
Formula: C₈H₃NO₅
MW: 193.12 g/mol
MDL Number: MFCD00005922
CAS Number: 5466-84-2
EINECS: 226-776-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR