Dicarbonylbis(triphenylphosphine)nickel, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Bis(triphenylphosphine)dicarbonylnickelBis(triphenylphosphine)nickel dicarbonyl

M-Clarity™ quality level = MQ100

SIAL213934-10GEA 152 GBP
SIAL213934-10G
Dicarbonylbis(triphenylphosphine)nickel, Sigma-Aldrich®
Dicarbonylbis(triphenylphosphine)nickel
Formula: C₃₈H₃₀NiO₂P₂
MW: 639.29 g/mol
MDL Number: MFCD00009591
CAS Number: 13007-90-4
EINECS: 235-851-9
UN: 3466
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR