D-(-)-Quinic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid

M-Clarity™ quality level = MQ200

SIAL138622-25GEA 96.5 GBP
SIAL138622-25G SIAL138622-100G
D-(-)-Quinic acid, Sigma-Aldrich®
D(-)-Quinic acid
Formula: C₇H₁₂O₆
MW: 192.17 g/mol
MDL Number: MFCD00003864
CAS Number: 77-95-2
EINECS: 201-072-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR