D(-)-Quinic acid

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid

MOLE28404581-100GEA 0 GBP
MOLE28404581-100G
D(-)-Quinic acid
D(-)-Quinic acid
Formula: C₇H₁₂O₆
MW: 192.17 g/mol
MDL Number: MFCD00003864
CAS Number: 77-95-2
EINECS: 201-072-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR