5-Bromo-2-furoic acid
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Bromofuran-2-carboxylic acid
, 5-Bromofuroic acid
MOLE41675121-25GEA
0
GBP
MOLE41675121-25G
5-Bromo-2-furoic acid
5-Bromo-2-furoic acid
Formula:
C₅H₃BrO₃ MW: 190.98 g/mol |
MDL Number:
MFCD00003239 CAS Number: 585-70-6 EINECS: 209-559-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...