5-Bromo-2-furoic acid

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromofuran-2-carboxylic acid5-Bromofuroic acid

FLUOF020339-1GEA 0 GBP
FLUOF020339-1G
5-Bromo-2-furoic acid
5-Bromo-2-furoic acid
Formula: C₅H₃BrO₃
MW: 190.98 g/mol
MDL Number: MFCD00003239
CAS Number: 585-70-6
EINECS: 209-559-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR