1-(4-tert-Butylbenzyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019236-1G 019236-100G 019236-25G 019236-5G
FLUO019236-1GEA 0 GBP
FLUO019236-1G FLUO019236-100G FLUO019236-25G FLUO019236-5G
1-(4-tert-Butylbenzyl)piperazine
1-(4-tert-Butylbenzyl)piperazine
Formula: C₁₅H₂₄N₂
MW: 232.37 g/mol
MDL Number: MFCD00082594
CAS Number: 956-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR