5-Methoxybenzofuran
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018772-1G
018772-5G
FLUO018772-1GEA
0
GBP
FLUO018772-1G
FLUO018772-5G
5-Methoxybenzofuran
5-Methoxybenzofuran
Formula:
C₉H₈O₂ MW: 148.16 g/mol |
MDL Number:
MFCD00079770 CAS Number: 13391-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...