2-Acetyl-5-nitrobenzofuran
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013109-1G
013109-5G
FLUO013109-1GEA
0
GBP
FLUO013109-1G
FLUO013109-5G
2-Acetyl-5-nitrobenzofuran
2-Acetyl-5-nitrobenzofuran
Formula:
C₁₀H₇NO₄ MW: 205.17 g/mol |
MDL Number:
MFCD00051634 CAS Number: 23136-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...