2-Bromo-1-ethylpyridinium tetrafluoroborate

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate2-Bromo-1-ethyl-pyridinium tetrafluoroborate

FLUO008541-5GEA 0 GBP
FLUO008541-5G
2-Bromo-1-ethylpyridinium tetrafluoroborate
2-Bromo-1-ethylpyridinium tetrafluoroborate
Formula: C₇H₉BBrF₄N
MW: 273.86 g/mol
MDL Number: MFCD00060046
CAS Number: 878-23-9
EINECS: 212-900-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR