Vitamin A palmitate

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Retinol palmitateRetinyl palmitateall-trans-Retinyl palmitate

EHERCA17923840EA 144 GBP
EHERCA17923840
Vitamin A palmitate
Vitamin A palmitate
Formula: C₃₆H₆₀O₂
MW: 524.87 g/mol
MDL Number: MFCD00019414
CAS Number: 79-81-2
EINECS: 201-228-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR