Vitamin A acetate
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
all-trans-Retinyl acetate
, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate
EHERCA17923820EA
232
GBP
EHERCA17923820
Vitamin A acetate
all-trans-Retinyl acetate
Formula:
C₂₂H₃₂O₂ MW: 328.49 g/mol |
MDL Number:
MFCD00019413 CAS Number: 127-47-9 EINECS: 204-844-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...