2-(Boc-amino)-3-methylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(N-Boc-Amino)-3-methylpyridine

PY-7971-1G PY-7971-5G
COBBPY-7971-1GEA 0 GBP
COBBPY-7971-1G COBBPY-7971-5G
2-(Boc-amino)-3-methylpyridine
2-(Boc-amino)-3-methylpyridine
Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
MDL Number: MFCD03425358
CAS Number: 138343-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR