2-Bromo-4-methoxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7397-1G PY-7397-5G
COBBPY-7397-1GEA 0 GBP
COBBPY-7397-1G COBBPY-7397-5G
2-Bromo-4-methoxypyridine
2-Bromo-4-methoxypyridine
Formula: C₆H₆BrNO
MDL Number: MFCD06254613
CAS Number: 89488-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR