2-Acetamido-5-chloro-4-methylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7185-1G PY-7185-5G
COBBPY-7185-1GEA 0 GBP
COBBPY-7185-1G COBBPY-7185-5G
2-Acetamido-5-chloro-4-methylpyridine
2-Acetamido-5-chloro-4-methylpyridine
Formula: C₈H₉ClN₂O
MDL Number: MFCD07375088
CAS Number: 148612-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR