2-(Boc-amino)-5-bromopyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-Boc-2-Amino-5-bromopyridine, 2-Amino-5-bromopyridine, 2-BOC protected
OT-8046-5G
OT-8046-25G
COBBOT-8046-5GEA
0
GBP
COBBOT-8046-5G
COBBOT-8046-25G
2-(Boc-amino)-5-bromopyridine
2-(Boc-amino)-5-bromopyridine
Formula:
C₁₀H₁₃BrN₂O₂ MW: 273.13 g/mol |
MDL Number:
MFCD06411381 CAS Number: 159451-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...