4-Bromosalicylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-2-hydroxybenzoic acid

OS-7530-100G OS-7530-5G OS-7530-25G OS-7530-1G
COBBOS-7530-100GEA 0 GBP
COBBOS-7530-100G COBBOS-7530-5G COBBOS-7530-25G COBBOS-7530-1G
4-Bromosalicylic acid
4-Bromosalicylic acid
Formula: C₇H₅BrO₃
MW: 217.02 g/mol
MDL Number: MFCD06797970
CAS Number: 1666-28-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR