4-Picolyl chloride hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Chloromethyl)pyridine hydrochloride, 4-(Chloromethyl)-pyridinium chloride

OS-7433-100G OS-7433-25G OS-7433-5G
COBBOS-7433-100GEA 0 GBP
COBBOS-7433-100G COBBOS-7433-25G COBBOS-7433-5G
4-Picolyl chloride hydrochloride
4-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
MDL Number: MFCD00012826
CAS Number: 1822-51-1
EINECS: 217-350-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR