5-Bromo-2-iodobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3199-5G HC-3199-25G HC-3199-1G
COBBHC-3199-5GEA 0 GBP
COBBHC-3199-5G COBBHC-3199-25G COBBHC-3199-1G
5-Bromo-2-iodobenzoic acid
5-Bromo-2-iodobenzoic acid
Formula: C₇H₄BrIO₂
MDL Number: MFCD00079717
CAS Number: 21740-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR