1-(4-Amino-2-fluorophenyl)-4-methylpiperazine

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoro-4-(4-methylpiperazino)aniline, 3-fluoro-4-(4-methyl-1-piperazinyl)benzenamine, 3-Fluoro-4-(4-methylpiperazin-1-yl)aniline

6R-1024.500 6R-1024.1 6R-1024.5 6R-1024.10
BIOT6R-1024.500EA 0 GBP
BIOT6R-1024.500 BIOT6R-1024.1 BIOT6R-1024.5 BIOT6R-1024.10
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine
Formula: C₁₁H₁₆FN₃
MW: 209.27 g/mol
MDL Number: MFCD05663793
CAS Number: 221198-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR