2-Chloro-3-formylpyridine

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloropyridine-3-carboxaldehyde, 2-Chloronicotinaldehyde, 2-Chloro-3-pyridinecarboxaldehyde

2L-376S.500 2L-376S.1 2L-376S.5 2L-376S.10
BIOT2L-376S.500EA 0 GBP
BIOT2L-376S.500 BIOT2L-376S.1 BIOT2L-376S.5 BIOT2L-376S.10
2-Chloro-3-formylpyridine
2-Chloro-3-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
MDL Number: MFCD01315308
CAS Number: 36404-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR