3-Hydroxy-2,4,6-triiodobenzoic acid ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
HTBA
B23355.06
B23355.14
B23355.06EA
44.2
GBP
B23355.06
B23355.14
3-Hydroxy-2,4,6-triiodobenzoic acid ≥97%
3-Hydroxy-2,4,6-triiodobenzoic acid
Purity: 97%
CAS No.: 53279-72-4
Molecular Formula: C7H3I3O3
Formula Weight: 515.81
Melting Point: 211-215°
Beilstein Reference: 2113494
Formula:
C₇H₃I₃O₃ MW: 515.81 g/mol Melting Pt: 211…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020305 CAS Number: 53279-72-4 EINECS: 258-457-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...