Fluorocyclohexane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010461-5G 010461-10G 010461-1G
FLUO010461-5GEA 0 GBP
FLUO010461-5G FLUO010461-10G FLUO010461-1G
Fluorocyclohexane
Fluorocyclohexane
Formula: C₆H₁₁F
MW: 102.15 g/mol
Boiling Pt: 98…100 °C (1013 hPa)
Density: 0,928 g/cm³ (19.9 °C)
MDL Number: MFCD00021285
CAS Number: 372-46-3
EINECS: 206-754-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR