Cyclopentanecarboxylic acid 98+%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

111490250 111491000
ACRO111490250EA 61.4 GBP
ACRO111490250 ACRO111491000
Cyclopentanecarboxylic acid 98+%
Cyclopentanecarboxylic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.05 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00001371
CAS Number: 3400-45-1
EINECS: 222-269-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4550 (20°C, 589 nm)
GC ≥98.0 %
Titration with NaOH 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR