Cyclopentanone

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

EHERC11833500EA 85 GBP
EHERC11833500
Cyclopentanone
Cyclopentanone
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –58 °C
Density: 0.941 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00001409
CAS Number: 120-92-3
EINECS: 204-435-9
UN: 2245
ADR: 3,III
Merck Index: 13,02771

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR