2-Quinolinol

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-QuinoloneQuinolin-2(1H)-one2-HydroxyquinolineCarbostyrilQuinolin-2-ol

MOLE64661238-5GEA 0 GBP
MOLE64661238-5G
2-Quinolinol
2-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 199…200 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006743
CAS Number: 59-31-4
EINECS: 200-420-6
Merck Index: 13,01835

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR