2,4-Dimethylthiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,4-Dimethylbenzene-1-thiol, 1,3-Dimethyl-4-mercaptobenzene, 4-Mercapto-m-xylene, 2,4-Dimethylbenzenethiol

001344-5G 001344-25G
FLUO001344-5GEA 0 GBP
FLUO001344-5G FLUO001344-25G
2,4-Dimethylthiophenol
2,4-Dimethylthiophenol
Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 207…208 °C (1013 hPa)
Density: 1.027 g/cm³ (20 °C)
MDL Number: MFCD00010019
CAS Number: 13616-82-5
EINECS: 237-100-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR