2,6-Dimethylthiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,3-Dimethyl-2-mercaptobenzene, 2-Mercapto-m-xylene, 2,6-Dimethylbenzene-1-thiol, 2,6-Dimethylbenzenethiol

001346-5G 001346-25G
FLUO001346-5GEA 0 GBP
FLUO001346-5G FLUO001346-25G
2,6-Dimethylthiophenol
2,6-Dimethylthiophenol
Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 76 °C (1013 hPa)
Density: 1.038 g/cm³ (25 °C)
MDL Number: MFCD00010021
CAS Number: 118-72-9
EINECS: 204-272-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR