3,4-Dimethylthiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,2-Dimethyl-4-mercaptobenzene, 4-Mercapto-o-xylene, 3,4-Dimethylbenzene-1-thiol, 3,4-Dimethylbenzenethiol

001347-5G 001347-25G
FLUO001347-5GEA 0 GBP
FLUO001347-5G FLUO001347-25G
3,4-Dimethylthiophenol
3,4-Dimethylthiophenol
Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 58…59 °C (1 mmHg)
Melting Pt: 54…56 °C
Density: 1.0256 g/cm³ (20 °C)
MDL Number: MFCD00010023
CAS Number: 18800-53-8
EINECS: 242-587-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR