2-Amino-3,5-dimethylbenzoic acid 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3,5-Dimethylanthranilic acid

L07003.03 L07003.06
L07003.03EA 64.6 GBP
L07003.03 L07003.06
2-Amino-3,5-dimethylbenzoic acid 98%
2-Amino-3,5-dimethylbenzoic acid

Purity: 98% CAS No.: 14438-32-5 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: 189-194° Beilstein Reference: 1101785

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 189…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00017099
CAS Number: 14438-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR