2-Hydroxyiminopropiophenone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Hydroxyiminopropiophenone, 2-Isonitrosopropiophenone, 1-Phenyl-1,2-propanedione-2-oxime, 1-Phenyl-1,2-propanedione 2-Oxime
I3502-10G
I3502-50G
SIALI3502-10GEA
79.8
GBP
SIALI3502-10G
SIALI3502-50G
2-Hydroxyiminopropiophenone, Sigma-Aldrich®
2-Hydroxyiminopropiophenone
Formula:
C₉H₉NO₂ MW: 163.18 g/mol |
MDL Number:
MFCD00002115 CAS Number: 119-51-7 EINECS: 204-329-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...