Galactitol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dulcitol

M-Clarity™ quality level = MQ200

D0256-100G D0256-10MG
SIALD0256-100GEA 152 GBP
SIALD0256-100G SIALD0256-10MG
Galactitol, Sigma-Aldrich®
Galactitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064288
CAS Number: 608-66-2
EINECS: 210-165-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR