L(+)-Citrulline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
C7629-100G
C7629-10MG
C7629-1G
C7629-1KG
C7629-25G
C7629-500G
C7629-5G
SIALC7629-100GEA
693
GBP
SIALC7629-100G
SIALC7629-10MG
SIALC7629-1G
SIALC7629-1KG
SIALC7629-25G
SIALC7629-500G
SIALC7629-5G
L(+)-Citrulline, Sigma-Aldrich®
L(+)-Citrulline
Formula:
C₆H₁₃N₃O₃ MW: 175.19 g/mol |
MDL Number:
MFCD00064397 CAS Number: 372-75-8 EINECS: 206-759-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...