N-Acetyl-L-proline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
A0783-1G
A0783-5G
SIALA0783-1GEA
104
GBP
SIALA0783-1G
SIALA0783-5G
N-Acetyl-L-proline, Sigma-Aldrich®
N-Acetyl-L-proline
Formula:
C₇H₁₁NO₃ MW: 157.169 g/mol |
MDL Number:
MFCD00020837 CAS Number: 68-95-1 EINECS: 276-259-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...