3-Chlorothiophene-2-carbonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF382.5 CFF382.10 CFF382.25 017408-5G 017408-25G 017408-1G
CHMPCFF382.5EA 0 GBP
CHMPCFF382.5 CHMPCFF382.10 CHMPCFF382.25 FLUO017408-5G FLUO017408-25G FLUO017408-1G
3-Chlorothiophene-2-carbonyl chloride
3-Chlorothiophene-2-carbonyl chloride
Formula: C₅H₂Cl₂OS
MDL Number: MFCD00068137
CAS Number: 86427-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR