5-Methyl-2-thiopheneboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Methylthiophene-2-boronic acid, (5-Methylthiophen-2-yl)boronic acid
011005-1G
011005-5G
FLUO011005-1GEA
0
GBP
FLUO011005-1G
FLUO011005-5G
5-Methyl-2-thiopheneboronic acid
5-Methyl-2-thiopheneboronic acid
Formula:
C₅H₇BO₂S MW: 141.99 g/mol |
MDL Number:
MFCD01318166 CAS Number: 162607-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...