4-(Ethylthio)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Ethylthiophenylboronic acid, 4-(Ethylthio)benzeneboronic acid
BB-2639-25G
BB-2639-1G
BB-2639-100G
BB-2639-5G
COBBBB-2639-25GEA
0
GBP
COBBBB-2639-25G
COBBBB-2639-1G
COBBBB-2639-100G
COBBBB-2639-5G
4-(Ethylthio)phenylboronic acid
4-(Ethylthio)phenylboronic acid
Formula:
C₈H₁₁BO₂S MW: 182.05 g/mol |
MDL Number:
MFCD01318146 CAS Number: 145349-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...