4-(Methylthio)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Methylthio)benzeneboronic acid, 4-Thioanisoleboronic acid

BB-2472-5G BB-2472-100G BB-2472-25G
COBBBB-2472-5GEA 0 GBP
COBBBB-2472-5G COBBBB-2472-100G COBBBB-2472-25G
4-(Methylthio)phenylboronic acid
4-(Methylthio)phenylboronic acid
Formula: C₇H₉BO₂S
MW: 168.02 g/mol
MDL Number: MFCD00093410
CAS Number: 98546-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR