4-Fluoro-7-nitrobenzofurazan

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Synonyms: NBD-F4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole

90046.EA 280 GBP
90046.
4-Fluoro-7-nitrobenzofurazan
4-Fluoro-7-nitrobenzofurazan
Formula: C₆H₂FN₃O₃
MW: 183.1 g/mol
MDL Number: MFCD00010196
CAS Number: 29270-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR