4-Chlorophenylurea ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(4-Chlorophenyl)urea

B25090.06 B25090.14
B25090.06EA 28.1 GBP
B25090.06 B25090.14
4-Chlorophenylurea ≥98%
4-Chlorophenylurea

Purity: 98% CAS No.: 140-38-5 Molecular Formula: C7H7ClN2O Formula Weight: 170.60 Melting Point: 204-205° Beilstein Reference: 908492

Formula: C₇H₇ClN₂O
MW: 170.6 g/mol
Melting Pt: 204…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00014788
CAS Number: 140-38-5
EINECS: 205-412-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR