Tetradecylbenzene, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
87204-500MGEA
143
GBP
87204-500MG
Tetradecylbenzene, Supelco®
Tetradecylbenzene
Formula:
C₂₀H₃₄ MW: 274.49 g/mol Boiling Pt: 354 °C (1013 hPa) Melting Pt: 16.1 °C Density: 0.8559 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00015080 CAS Number: 1459-10-5 EINECS: 215-950-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...