1-tert-Butyl-4-nitrobenzene ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-(tert-Butyl)-4-nitrobenzene

L06067.09 L06067.18 L06067.04
L06067.09EA 60.9 GBP
L06067.09 L06067.18 L06067.04
1-tert-Butyl-4-nitrobenzene ≥98%
1-tert-Butyl-4-nitrobenzene

Purity: 98% CAS No.: 3282-56-2 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Refractive Index: 1.5340 Boiling Point: 265-267° MDL No.: MFCD00041253

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Boiling Pt: 265…267 °C
Density: 1.0586
Storage Temperature: Ambient
MDL Number: MFCD00041253
CAS Number: 3282-56-2
EINECS: 221-922-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR