4-Fluoro-3-methylanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

006276-5G 006276-25G
FLUO006276-5GEA 0 GBP
FLUO006276-5G FLUO006276-25G
4-Fluoro-3-methylanisole
4-Fluoro-3-methylanisole
Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 23 °C (1013 hPa)
Density: 1,07 g/cm³ (20 °C)
MDL Number: MFCD00042293
CAS Number: 2338-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR